Bossheads for frames, 90°
Supplier: BOCHEM
Jump to:
About this item
For rods up to 13 mm.
- Angle: 90°
- Heart screws
2 Options Available Below
Product Details & Documents
For rods up to 13 mm.
- Angle: 90°
- Heart screws
For rods up to 13 mm.
2 Options Available Below
For rods up to 13 mm.