VWR®, Disposable Weighing Boats
PS, diamond.
- Wide, flat bottom avoids tipping
- Temperature resistant up to +70 °C
These flexible boats feature smooth surfaces to provide accurate pour-out with minimal sample loss. They are ideal for use with microwaves and moisture analysers.
: They are produced and packed in plastic bags in a cleanroom.