



Boron trifluoride - methanol complex 14% in methanol, Sigma-Aldrich®
Supplier: SIGMAALDRICH
Certificates
About this item
CAS Number:373-57-9
:16045-88-8
Formula:CH₄BF₃O
MDL Number:MFCD00071635
:99.85 g/mol
EINECS:206-766-4
UN:3286
ADR:3,II
:GHS02
:GHS06
:GHS05
:GHS08