Order Entry
China
ContactUsLinkComponent
 
Potassium iodo mercurate(II) reagent, solution B (sodium hydroxide 20% aqueous solution) Reag. Ph. Eur. 1071600 for determination of ammonia and ammonium salts
Catalog # VWRC87710.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium iodo mercurate(II) reagent, solution B (sodium hydroxide 20% aqueous solution) Reag. Ph. Eur. 1071600 for determination of ammonia and ammonium salts
Catalog # VWRC87710.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

 :
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
MDL Number:MFCD00003548