Order Entry
China
ContactUsLinkComponent
Gadolinium trinitrate hexahydrate 99.9% (trace metals basis)
 
 
Gadolinium trinitrate hexahydrate 99.9% (trace metals basis)
  ACRO316760250
 :  
undefined
 
 
Gadolinium trinitrate hexahydrate 99.9% (trace metals basis)
  ACRO316760250
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White Crystalline powder or crystals
  • Total conc. of imp.as REO:
    ≤0.1 %
  • Gadolinium (Gd):
    34 to 36 %

 

 

 :
 :451.36
 :91 °C
 :2,4 g/cm³ (20 °C)
 :Ambient
 :233-437-2
 :1477
 :5.1,III
 :GHS05
 :GHS03
 :19598-90-4
 :GdN₃O₉·6H₂O
 :MFCD00149728