Strips of 8 PCR tubes and cap strips, Corning® Thermowell® GOLD
证书
关于此项目
Tube strips consist of eight 0,2 ml thin wall PP tubes connected together. Dual connectors between adjacent tubes eliminate inadvertent breakage during sample handling. Tube strips are designed for precise fit in thermal cyclers to optimise heat transfer. Thermowell® GOLD cap strips are sold separately. Tube strips are tested and certified to be free of DNase/RNase contamination and are autoclavable to 121 °C.
The optically clear, 8-well flat cap strips feature an ultra-clear surface ideal for real-time PCR and fluorescent and luminescent applications.
Ordering information: Assorted pack includes the following colours: blue, green, orange, red, violet, and yellow.
规格
- Catalog No:
- CORN6542
- CORN3742
- CORN6547
- CORN3743
- CORN3740
- CORN3741
- CORN3748
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- 环境认证:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa