Magnesium chloride hexahydrate USP, Multi-Compendial, J.T.Baker®
Catalog # BAKR2449-40
Supplier: Avantor
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Poly pail
- Pk:9,2 kg
- Seq:4152
Specifications
Specification Test Results
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - pH (1 in 20):4.5 - 7.0
- USP - Insoluble Matter:≤ 0.005 %
- USP - Sulfate (SO₄):≤ 0.005 %
- USP - Barium (Ba):Passes Test
- USP - Calcium (Ca):≤ 0.01 %
- USP - Potassium (K):Passes Test
- USP - Assay:98.0 - 101.0 %
- USP - Endotoxin Concentration (EU/g):≤ 2.5
- EP/BP - Assay:98.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Bromide (Br):≤ 500 ppm
- EP/BP - Sulfate (SO₄):≤ 100 ppm
- EP/BP - Calcium (Ca):≤ 0.1 %
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Water (H₂O):51.0 - 55.0 %
Specification Test Results
About this item
:
:203.3 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:117 °C
Density:1.57 g/cm³ (25 °C)
:Management of Change (MOC) category = HR
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7791-18-6
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781