Specifications
- Description:EasySep™ direct human T cell isolation kit, 100 ml whole blood processed
- Seq:01
- Pk:1 KIT
Specifications
About this item
Immunomagnetic isolation of T cells directly from whole blood
- >99,9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
- Up to 97% purity of isolated cells
- Fast, easy-to-use and column-free
- Isolated cells are untouched
This cell separation kit can be used to isolate highly purified T cells directly from human whole blood by immunomagnetic negative selection.
This kit targets non-T cells for removal with antibodies recognising specific surface markers. Unwanted cells are labelled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet. Desired cells are simply collected into a new tube and can directly be used for functional assays, flow cytometry or DNA/RNA extraction.