Order Entry
China
ContactUsLinkComponent
EasySep™ Direct Human NK Cell Isolation Kit
EasySep™ Direct Human NK Cell Isolation Kit
Catalog # STMC19665
undefined
EasySep™ Direct Human NK Cell Isolation Kit
Catalog # STMC19665
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    EasySep™ direct human NK cell isolation kit, 100 ml whole blood processed
  • Pk:
    1 KIT
Specifications
About this item
Immunomagnetic isolation of NK cells directly from whole blood
  • >99,9% red blood cell (RBC) depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 84% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ Direct Human NK Cell Isolation Kit is designed to isolate NK cells from human whole blood by immunomagnetic negative selection. The EasySep™ Direct procedure involves labeling RBCs, platelets and unwanted cells present in human whole blood with antibody complexes and magnetic particles. The magnetically labelled RBCs, platelets and cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').