Specifications
- Description:EasySep™ mouse Naïve CD4+ T cell isolation kit, 1×10⁹ cells
- Seq:01
- Pk:1 KIT
Specifications
About this item
Immunomagnetic isolation of untouched mouse naive CD4+ T cells
- Fast, easy-to-use and column-free
- Up to 95% purity
- Untouched, viable cells
The EasySep™ Mouse Naïve CD4+ T Cell Isolation Kit is designed to isolate naïve CD4+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-naïve CD4+ T cells , featuring RapidSphere™ technology.