Specifications
- Description:EasySep™ human Naïve CD4+ T cell Isolation kit II, 1×10⁹ cells
- Seq:01
- Pk:1 KIT
Specifications
About this item
Immunomagnetic isolation of untouched human naïve CD4+ T cells.
- Fast, easy-to-use and column-free
- Up to 98% purity
- Untouched, viable cells
The EasySep™ Human Naïve CD4+ T Cell Isolation Kit II is designed to isolate naïve CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This kit replaces the EasySep™ Human Naïve CD4+ T Cell Enrichment Kit (STMC19155) and EasySep™ Human Naïve CD4+ T Cell Isolation Kit (STMC19555) for even faster cell isolations.