Order Entry
China
ContactUsLinkComponent
EasySep™ Human CD14 Positive Selection Kit II
  STMC17858
undefined
EasySep™ Human CD14 Positive Selection Kit II
  STMC17858
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    EasySep™ human CD14 positive selection kit II, 1×10⁹ cells
  • Seq:
    01
  • Pk:
    1 KIT
Immunomagnetic positive selection of human CD14+ cells (e.g. monocytes).
  • Fast and easy-to-use
  • Up to 97% purity
  • No columns required
The EasySep™ Human CD14 Positive Selection Kit II is designed to isolate highly purified CD14+ cells from fresh or previously frozen human peripheral blood mononuclear cells (PBMCs). Desired cells are targeted with Tetrameric Antibody Complexes recognising CD14 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD14 antigen is strongly expressed on monocytes and macrophages and weakly on granulocytes. It is also expressed on most tissue macrophages.
This product replaces the EasySep™ Human CD14 Positive Selection Kit (STMC18058), for even faster cell isolations and is compatible with all EasySep™ magnets.
 : Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).