Order Entry
China
ContactUsLinkComponent
Silver nitrate 0.25 mol/l (0.25 N) in aqueous solution Reag. Ph. Eur. 1078301
Silver nitrate 0.25 mol/l (0.25 N) in aqueous solution Reag. Ph. Eur. 1078301
Catalog # VWRC87924.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Silver nitrate 0.25 mol/l (0.25 N) in aqueous solution Reag. Ph. Eur. 1078301
Catalog # VWRC87924.290
Supplier:  VWR Chemicals
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Performance test:
    Passes test

Specification Test Results

About this item

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
 :
 :169.87 g/mol
Density:1.033 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-853-9
UN:3264
ADR:8,III
 :GHS05
 :GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414