Sodium hydroxide solution 10.0 N
Poly bottle, 1 L
Catalog # BDHBBDH7247-1
Supplier: BDH US PRODUCED
Sodium hydroxide solution 10.0 N
Catalog # BDHBBDH7247-1
Supplier: BDH US PRODUCED
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
APHA. Made with deionised. Suitable as a titrant.
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
Specifications
Specification Test Results
- Appearance/colour:Clear liquid
- N.I.S.T. traceable to SRM 84:Yes
- Normality:9.950 - 10.050 N
- Shelf-life:2 years
Specifications
- Pack type:Poly bottle
- Pk:1 L



