

2-Propanol ≥99.9% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Catalog # HONEAH323-4
Supplier: HONEYWELL SPECIALTY CHEMICALS
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Carbonyl Compounds:0.002 % max.
- Water by Karl Fischer Titration:0.06 % max.
- Color (APHA):10 max.
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.3766 - 1.3778
- Residue:3 mg/L max.
- Titrable Acid or Base:0.1 µeq/g max.
- UV Absorbance @ 225 nm:0.16 AU max.
- UV Absorbance @ 250 nm:0.02 AU max.
- UV Cutoff:205 nm max.
Specification Test Results
About this item
:
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:12.0 %(V)
Melting Pt:–88 °C
Density:0.785 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS02
:GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674