



Crystal violet, pure indicator
Catalog # ACRO212120250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Dark green Crystalline powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Sensitivity:(as indicator) Passes test
Specification Test Results
About this item
:
:407.99 g/mol
Merck Index:13,04406
Melting Pt:173 °C (dec.)
Density:1,19 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-953-6
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS05
:GHS09
CAS Number:548-62-9
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750