Order Entry
China
ContactUsLinkComponent
VWR®, Organic reference standard, Aroclor 1260 (PCB-mixture, 60% chlorinated) 35 µg/ml in isooctane
VWR®, Organic reference standard, Aroclor 1260 (PCB-mixture, 60% chlorinated) 35 µg/ml in isooctane
Catalog # VWRC122732H
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Organic reference standard, Aroclor 1260 (PCB-mixture, 60% chlorinated) 35 µg/ml in isooctane
Catalog # VWRC122732H
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    1 mL
  • Description:
    Aroclor 1260 (PCB-mixture, 60% chlorinated) 35 µg/ml in isooctane

Specifications

About this item