Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phenolphthalein
Phenolphthalein 0.5% in aqueous solution
Phenolphthalein 0.5% in aqueous solution
Phenolphthalein 0.5% in aqueous solution
HACH16232
:
HACH
:
Phenolphthalein 0.5% in aqueous solution
HACH16232
:
HACH
:
Pack type:
Glass bottle
Pk:
100 mL
Seq:
2440
:
:
318.33 g/mol
:
Ambient
:
201-004-7
:
77-09-8
:
C₂₀H₁₄O₄
:
MFCD00005913