Congo red indicator grade
: Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), CI 22120, Direct Red 28
:573-58-0
:
:C₃₂H₂₂N₆Na₂O₆S₂
:MFCD00004028
:696.67 g/mol
:12,02562
:>360 °C
:209-358-4
:GHS08
:Ambient
- Environmentally Preferable:
- Appearance:Dark red to red-brown Powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Solubility:(0.1 % in water) Clear
- Visual transition interval:from pH 3.0 (violet) to pH 5.2 (red-orange)