ELISA Carbonate Coating Buffer
Catalog # TFBRCB01100
Supplier: THERMO FISHER BRAND
New Product
CAS Number:
Specifications
- Pk:1
- Description:ELISA Carbonate coating buffer, 100 ml
- Environmentally Preferable:
- Storage temperature:Store at 2...8 °C
- For use with:ELISA
- Physical form:Concentrate
- pH value:9,4
Specifications
About this item
ELISA Carbonate coating buffer is intended for use as a plate coating buffer for Invitrogen Antibody Pairs or when developing a sandwich ELISA.
- The buffer is a 50 mM carbonate buffer, pH 9,4
For use as a plate coating buffer for invitrogen antibody pairs or when developing a sandwich ELISA.