Industrial Microscope, BX53M
Designed with modularity in mind, the BX3M series provide versatility for a wide variety of materials science and industrial applications. With improved integration with PRECiV software, the BX53M provides a seamless workflow for standard microscopy and digital imaging users from observation to report creation.
- Simple illuminator; intuitive microscope controls
- Find the focus quickly; consistent illumination
- Easy and ergonomic operation; easy restore microscope settings
- Basic measurement functions
- Create all-in-focus images; easily move the stage for panorama
- Capture both bright and dark areas; adaptable to suit observational and analysis preferences
- Accommodates a wide range of samples
The BX53M simplifies complex microscopy tasks through its well-designed and easy-to-use controls. Users can get the most out of the microscope without the need for extensive training. The easy, comfortable operation of the BX53M also improves reproducibility by minimising human error.
The BX53M maintains the traditional contrast methods of conventional microscopy, such as brightfield, darkfield, polarised light, and differential interference contrast. As new materials are developed, many of the difficulties associated with detecting defects using standard contrast methods can be solved using advanced microscopy techniques for more accurate and reliable inspections. New illumination techniques and options for image acquisition within PRECiV image analysis software give users more choices of how to evaluate their samples and document findings.
- Model:BX53M
- Description:Industrial trinocular microscope, 25 mm stroke, 1 μm minimum graduation, with upper limit stopper, torque adjustment for coarse handle
- Cat. No.:OLYMBX53M
- Optical design:UIS2 optical system (infinity-corrected)