Order Entry
China
Orders LinkContactUsLinkComponent
PolarSafe™ Cooling Blocks
PolarSafe™ Cooling Blocks
Aluminum cooling block, 15-well for 1,5/2,0 ml tubes, 15, Blue
Catalog # ARGT63615-01
Supplier:   Argos Technologies
PolarSafe™ Cooling Blocks
Catalog # ARGT63615-01
Supplier:  Argos Technologies
PolarSafe™ Cooling Blocks
Aluminum cooling block, 15-well for 1,5/2,0 ml tubes, 15, Blue
Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Quit running to the freezer for more ice.
  • Cool or heat your samples quickly—blocks adjust to desired temperature within minutes
  • Eliminate uneven cooling
  • Keep samples upright and organized while controlling temperature
  • Contoured wells hold tubes and vials snugly for excellent temperature convection
Place blocks on ice, dry ice, liquid nitrogen, or in a water bath. These anodised aluminum blocks can keep samples at or near 1 °C for up to 8 hours in properly packed ice, or for 3 to 7 hours in dry ice. Choose the blocks that meet your application needs — configurations are available for 1,5-/2,0-ml microcentrifuge tubes and cryogenic vials, 0,5-ml tubes, and 96-well or 364-well plates.
Specifications
  • No. of Holes:
    15
  • Colour:
    Blue
  • Description:
    Aluminum cooling block, 15-well for 1,5/2,0 ml tubes
  • Seq:
    01