Order Entry
China
ContactUsLinkComponent
Ammonium heptamolybdate 10% in aqueous solution Reag. Ph. Eur. 1005702
Ammonium heptamolybdate 10% in aqueous solution Reag. Ph. Eur. 1005702
Catalog # VWRC87770.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium heptamolybdate 10% in aqueous solution Reag. Ph. Eur. 1005702
Catalog # VWRC87770.290
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :1235.86 g/mol
EINECS:234-722-4
CAS Number:12054-85-2
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059