Order Entry
China
ContactUsLinkComponent
 
PHCbi Low Temperature Incubator, 154 L
  SANYMIR-154L-PC
 :  SANYO
PHCbi Low Temperature Incubator, 154 L
  SANYMIR-154L-PC
 :  SANYO

 

  • Weight:
    78 kg
  • W×D×H ext.:
    700×580×1018 mm
  • Capacity:
    123 L
  • Description:
    Low temperature constant temperature incubator
  • No. of shelves:
    3
  • Temperature setting accuracy:
    −10...+60 °C
  • Pk:
    1
  • W×D×H int.:
    620×368×5555 mm

 

 

PHC low temperature incubator provides the most stable culture environment.

  • Precise and stable temperature environment
  • PID control, low-temperature incubator equipped with high-precision PID temperature control system and compressor switch system, can achieve temperature fluctuation ≤±0,2 °C @ box 50 °C, ambient temperature 20 °C, ≤±1,5 °C @ box 5 °C, ambient temperature 20 °C
  • LCD graphic display makes operation easier, and can achieve 12 programs, 98 steps of setting
  • The internal fan does not blow directly to the sample to reduce the evaporation of the sample culture medium
  • Optional lighting component, located on the top of the incubator, the illumination can reach 3000 lux

Widely used in:
Stability tests of drugs, packaging materials, reagents, etc. in medical and pharmaceutical industries.
Stability tests of foods, food preservation tests, and quality management of frozen dairy products in the food industry.
Specific gravity and viscosity determination of adhesives in the chemical industry, and adhesive preservation tests. Acid-base tests of coatings, stability tests of soaps and detergents, and stability tests of powdered and liquid samples. Plastic hardening tests, and oil deterioration tests over the years in the agronomy field. Drainage BOD detection. Fungus culture and preservation. Rice saccharification tests. Battery charging and discharging reagents, battery cycle tests, and battery aging characteristics tests in the electronics field. Temperature characteristics tests of semiconductors, and preservation of semiconductor inductive materials. Polymer material hardening tests. Environmental tests in the metrology department. Constant value operation of morphological memory alloy material tests in the mechanical metal field. Adhesion durability tests of labels and adhesive strips, printing adhesives, organic solvent preservation, and ink preservation tests.