CDK turbomolecular pumping systems are completely oil-free. It incorporate an oil-free SST turbomolecular pump using dry-running, solid-lubricated ceramic bearings. The backing pump is an oil-free diaphragm vacuum pump.
- Unique, built-in automatic shut-off device for the backing pump
- User friendly
- Automatic shut-off device for the backing pump
- Lightweight, compact and portable
If the pump system works continually at the ultimate pressure, and if the connected vacuum system has a low real and virtual leak rate (wall degassing), then it is possible that the backing pump will remain switched off for hours.
The CDK systems have all components housed inside a casing, making them very compact and portable. The components include turbomolecular pump, backing pump, controller, and connecting tubing. A simple On/Off button makes operation simple. CDK systems employ three-stage chemical duty diaphragm pump as the backing pump.
- Catalog No:
- ILMV101241
- ILMV101243
- Model:
- CDK 180 UHV
- CDK 280 UHV
- Power connection:
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- Weight:
- 16,7 kg
- 16,7 kg