- Pk:5 mg
- Description:Click-IT™ AHA (L-Azidohomoalanine)
- Storage conditions:Store in freezer
- Shipping conditions:RT
- Storage temperature:−5...−30 °C
Click-iT® AHA (L-azidohomoalaine) provides a fast, sensitive, non-toxic and most importantly non-radioactive alternative to the traditional radioactive technique, 35S-methionine for the detection of nascent protein. AHA is an amino acid analog that contains a very small modification, specifically an azido moiety that can be fed to cultured cells and incorporated into proteins during active protein synthesis.
- Less hazardous
- AHA product type
- Biotin-based, fluorescent detection method
- Metabolic labeling
- Proteins labeling target
Detection utilises the chemoselective ligation or 'click' reaction between an azide and an alkyne where the azido modified protein is detected with one of the Click-iT® protein analysis detection kits containing either TAMRA, Dapoxyl®, or biotin alkyne. Detection sensitivity with these reagents in 1-D gels and western blots is in the low femtomole range and compatible with downstream LC-MS⁄MS and MALDI MS analysis or Multiplexed Proteomics® reagents for differential analyses of newly synthesised proteins together with total glycoproteins, total phosphoproteins or total protein.