Order Entry
China
ContactUsLinkComponent
Disodium hydrogen phosphate 98.0-100.5% (dried basis) USP, J.T.Baker®
Disodium hydrogen phosphate 98.0-100.5% (dried basis) USP, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Disodium hydrogen phosphate 98.0-100.5% (dried basis) USP, J.T.Baker®
Supplier:  Avantor
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • GMP Manufactured Product:
    98.0 - 100.5%
  • Meets U.S.P Requirements:
    ≤3 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.06%
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Assay (Na₂HPO₄) (dried basis):
    98.0 - 100.5 %
  • USP - Arsenic (As):
    ≤ 3 ppm
  • USP - Chloride (Cl):
    ≤ 0.06 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Insoluble Matter:
    ≤ 0.2 %
  • USP - Loss on Drying at 130°C:
    ≤ 5.0 %
  • USP - Sulfate (SO₄):
    ≤ 0.2 %
  • (Suitable for Buffer Solutions):
    ≤0.005%
  • No Class 1,2,3 or other solvents are used or produced in the:
    ≤0.2%
  • manufacturing or purification of the product.:
    ≤4 mg/kg
  • Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
    ≤5.0%

Specification Test Results

Related Information