Ammonium iron (III) citrate, red brown, powder FCC, J.T.Baker®
BAKR1980-01
: Avantor
:
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Bulk Food Chemical:16.5 - 18.5 %
- FCC - Assay (as Fe):16.5 - 18.5 %
- Identification:Passes Test
- Ferric Citrate:Passes Test
- Lead (Pb):≤2 mg/kg
- Mercury (Hg):≤1 mg/kg
- Oxalate (C₂O₄):Passes Test
- Sulfate (SO₄):≤0.3 %
- Tartrate:Passes Test
:
:197 °C (1013 hPa)
:1.8 g/cm³ (20 °C)
:214-686-6
:1185-57-5
:C₆H₅+4y)Fe(x)N(y)O₇
:MFCD00013099