
Citric acid 99.5-100.5%, powder USP, Multi-Compendial, GMP, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:77-92-9
:
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669
:192.13 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149…151 °C
Density:1.67 g/cm³
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- GMP Manufactured Product:99.5 - 100.5%
- Meets B.P. Chemical Specifications:Passes Test
- Meets E.P. Chemical Specifications:Passes Test
- Meets J.P. Chemical Specifications:Passes Test
- Meets U.S.P Requirements:≤0.1%
- Preserve in Tight Containers:≤0.036%
- CAUTION: For Manufacturing, processing or repackaging:≤0.015%
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis):99.5 - 100.5 %
- USP - Clarity of Solution:Passes Test
- USP - Color of Solution:Passes Test
- USP - Identification:Passes Test
- USP - Residue on Ignition:≤ 0.1 %
- USP - Oxalic Acid:≤ 0.036 %
- USP - Sulfate (SO₄):≤ 0.015 %
- USP - Readily Carbonizable Substances:Passes Test
- USP - Identification C: Water (H₂O):≤ 1.0 %
- EP/BP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis):99.5 - 100.5 %
- EP/BP - Identification B:Passes Test
- EP/BP - Identification E:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Readily Carbonizable Substances:Passes Test
- EP/BP - Oxalic Acid:≤ 360 ppm
- EP/BP - Sulfate (SO₄):≤ 150 ppm
- EP/BP - Water (H₂O):≤ 1.0 %
- EP/BP - Ash (sulfated):≤ 0.1 %
- EP/BP - Endotoxin Concentration, <0.5 IU/mg:Passes Test
- JP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis):99.5 - 100.5 %
- JP - Clarity and Color of Solution:Passes Test
- JP - Identification:Passes Test
- JP - Sulfate (SO₄):≤ 150 ppm
- JP - Oxalic Acid:≤ 360 ppm
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Readily Carbonizable Substances:Passes Test
- JP - Residue on Ignition:≤ 0.10 %
- JP - Water (H₂O):≤ 1.0 %
- E. coli:Negative
- P. aureginosa:Negative