Anti-Casp3 (Asp175) cleaved Rabbit Monoclonal Antibody [clone: 5A1E]
Catalog # CSTA9664S
Supplier: CELL SIGNALING TECHNOLOGIES
Specifications
- Antibody type:Primary
- Antigen symbol:Casp3 (Asp175) cleaved
- Clonality:Monoclonal
- Clone:5A1E
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Casp3 (Asp175) cleaved
Clonality: Monoclonal
Clone: 5A1E
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey