Order Entry
China
ContactUsLinkComponent
Anti-p38-alpha MAPK (Thr180/Tyr182) phosphate Rabbit Monoclonal Antibody [clone: D3F9]
Anti-p38-alpha MAPK (Thr180/Tyr182) phosphate Rabbit Monoclonal Antibody [clone: D3F9]
  CSTA4511S
Anti-p38-alpha MAPK (Thr180/Tyr182) phosphate Rabbit Monoclonal Antibody [clone: D3F9]
  CSTA4511S

 

  • Antibody type:
    Primary
  • Antigen symbol:
    p38-alpha MAPK (Thr180/Tyr182) phosphate
  • Clonality:
    Monoclonal
  • Clone:
    D3F9
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    S. cerevisiae,
    Pig,
    Monkey,
    Human,
    Mink,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    02
  • Antigen synonyms:
    p38-delta MAPK (Thr180/Tyr182) phosphate|p38-alpha MAPK (Thr180/Tyr182) phosphate|p38-gamma MAPK (Thr183/Tyr185) phosphate|p38-beta MAPK (Thr180/Tyr182) phosphate
  • Sequence:
    LARHTDDEMtGyVAC
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Pk:
    200 µl

 

 

Type: Primary
Antigen: p38-alpha MAPK (Thr180/Tyr182) phosphate
Clonality: Monoclonal
Clone: D3F9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey;Mink;Pig;S. cerevisiae