Anti-p21 Waf1/Cip1 Rabbit Monoclonal Antibody [clone: 12D1]
Catalog # CSTA2947S
Supplier: CELL SIGNALING TECHNOLOGIES
Specifications
- Antibody type:Primary
- Antigen symbol:p21 Waf1/Cip1
- Clonality:Monoclonal
- Clone:12D1
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Western blot:Yes
- Seq:02
- Sequence:CPRSGEQAEGSPGGPGDSQGRKRRQT
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: p21 Waf1/Cip1
Clonality: Monoclonal
Clone: 12D1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Monkey