Order Entry
China
ContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) substrate system, liquid for ELISA, Sigma-Aldrich®

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
 :
 :240.35 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:166…170 °C
Storage Temperature:Ambient
EINECS:259-364-6
CAS Number:54827-17-7
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748