Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L), Sigma-Aldrich®
Catalog # SIALS7899-100ML
Supplier: SIGMAALDRICH
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Description:Sodium acetate buffer solution pH 5,2 (3 mol/l)
- M-Clarity quality level:MQ200