Order Entry
China
ContactUsLinkComponent
Hexane (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
Flammable
Health
Toxic
Environment
Hexane (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
Catalog # VWRC83992.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Toxic
Environment
Hexane (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
Catalog # VWRC83992.320
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 45.0 %
  • Acidity:
    Max. 0.001 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0.01 %
  • Transmittance (210 nm):
    Min. 60 %
  • Transmittance (220 nm):
    Min. 80 %
  • Transmittance (230 nm):
    Min. 95 %
  • Transmittance (240 nm):
    Min. 98 %
  • Transmittance (250 nm):
    Min. 99 %

Specification Test Results

About this item

 :
 :86.18 g/mol
Merck Index:13,04712
Boiling Pt:68…70 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
 :GHS02
 :GHS08
 :GHS06
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311