

Acetonitrile ≥99.95%, HiPerSolv CHROMANORM® ULTRA for LC-MS, suitable for UPLC/UHPLC-MS instruments
:75-05-8
:
:H₃CCN
:MFCD00001878
:41.05 g/mol
:13,00071
:81.6 °C (1013 hPa)
:–45,7 °C
:0.786 g/cm³ (20 °C)
:200-835-2
:1648
:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
:2 °C (closed cup)
:01-2119471307-38
:GHS02
:GHS07
:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Assay (on anhydrous substance):Min. 99.95 %
- Acidity:Max. 0.0001 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Colouration:Max. 5 APHA
- Density (20/4):0.781 --> 0.784
- Density (20/20):0.782 --> 0.785
- n 20/D:1.343 --> 1.345
- Evaporation residue:Max. 1 ppm
- Water:Max. 100 ppm
- Transmittance (193 nm):Min. 60 %
- Transmittance (195 nm):Min. 90 %
- Transmittance (200 nm):Min. 98 %
- Transmittance (210 nm):Min. 98 %
- Transmittance (215 nm):Min. 99 %
- Transmittance (220 nm):Min. 99 %
- Transmittance (230 nm):Min. 99 %
- Fluorescence (as quinine) (254/450 nm):Max. 1 ppb
- Fluorescence (as quinine) (365 nm):Max. 0.5 ppb
- Gradient (210 nm):Max. 1 mAU
- Gradient (254 nm):Max. 1 mAU
- MS-ESI+ (as Reserpine):Max. 2 ppb
- MS-ESI- (as 4-Nitrophenol):Max. 20 ppb
- MS-APCI- (as 4-Nitrophenol):Max. 20 ppb
- Al (Aluminium):Max. 20 ppb
- Ba (Barium):Max. 20 ppb
- Ca (Calcium):Max. 20 ppb
- Cd (Cadmium):Max. 20 ppb
- Co (Cobalt):Max. 20 ppb
- Cu (Copper):Max. 20 ppb
- Fe (Iron):Max. 20 ppb
- K (Potassium):Max. 20 ppb
- Mg (Magnesium):Max. 20 ppb
- Mn (Manganese):Max. 20 ppb
- Na (Sodium) (at filling):Max. 20 ppb
- Ni (Nickel):Max. 20 ppb
- Pb (Lead):Max. 20 ppb
- Zn (Zinc):Max. 20 ppb
- Filtered through 0.2 μm:Confirmed