
10-Undecenal 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-45-8
:
Formula:C₁₁H₂₀O
MDL Number:MFCD00007032
:168.28 g/mol
Boiling Pt:240 °C (1013 hPa)
Melting Pt:7 °C
Density:0.845 g/cm³
Flash Pt:76 °C
EINECS:203-973-1
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4400 to 1.4460 (20°C, 589 nm)
- GC:≥96.0 %
- Residue after evaporation:≤0.002 %
- Sulfur dioxide (SO2):≤2.5 ppm
- Hydrogen sulfide (H2S):≤1.5 ppm