

(Methoxymethyl)triphenylphosphonium chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4009-98-7
:
Formula:C₂₀H₂₀ClOP
MDL Number:MFCD00011800
:342.8 g/mol
Boiling Pt:413 °C
Melting Pt:195…197 °C
Flash Pt:>250 °C
Storage Temperature:Ambient
EINECS:223-664-5
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to almost white Powder or crystals
- Infrared spectrum:Conforms
- Water:≤0.5 %
- HPLC:≥97.5 %