Order Entry
China
ContactUsLinkComponent
2,3-Butanediol mixture of racemic and meso forms 98% tech.
2,3-Butanediol mixture of racemic and meso forms 98% tech.
Certificates
undefined
2,3-Butanediol mixture of racemic and meso forms 98% tech.

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4330 to 1.4370 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Absorption Ratio P-15/P+15:
    0.98 to 1.20
  • BSC Certification:
    Passes

Specification Test Results

Related Information