Order Entry
China
Orders LinkContactUsLinkComponent
Funnel holders
 :   BRAND
Funnel holders
 :  BRAND
Funnel holders
Funnel holders
PP, white.
To support 1 or 2 funnels with top outer-Ø of 50 to 120 mm. Easily mounted on supports rods with an outer-Ø 8 to 14 mm with a locking nut.
Product Family Options
  • Description
    For
  • Funnel holder for a funnel
    1 funnel
    1 * 5 件
    1 * 5 件
  • 2 funnels
    1 * 5 件
    1 * 5 件