Order Entry
China
ContactUsLinkComponent
ID Screen® Infectious Bronchitis Indirect 2.0 ELISA Kit
ID Screen® Infectious Bronchitis Indirect 2.0 ELISA Kit
Catalog # XXXXIBVARSV2-5P
Supplier:  OT CHEMICALS
CAS Number:  
ID Screen® Infectious Bronchitis Indirect 2.0 ELISA Kit
Catalog # XXXXIBVARSV2-5P
Supplier:  OT CHEMICALS
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Infectious bronchitis indirect 2.0 ELISA kit, 5 plates, 12×8-well strips
  • Tests per kit:
    480 reactions

Specifications

About this item

Indirect ELISA for the detection of antibodies against IBV in chicken serum, plasma or egg yolk samples.

  • Allows for improved detection of challenge in vaccinated flocks, especially by variant strains
  • High specificity and very low background (no risk of false positive results)
  • May be used with egg yolk samples which are easier to collect and cause less stress to the layers
  • Positive and negative reference sera available as internal controls
  • Comprehensive data analysis with the ID Soft™ program

Highly specific and with very low background, the ID Screen® infectious bronchitis indirect ELISA kit detects all IBV variants, including QX and 793B strains. It may be used to detect infection or to quantify IBV antibody levels in vaccinated birds.

Includes the same convenient protocol (buffers, incubation times and temperatures) as for most ID Screen® poultry kits with short incubation times: results in 75 minutes (30-30-15 protocol). Validated for use with serum transport cards, allowing for practical and safe shipment of blood or serum samples to the lab.

Method: Indirect ELISA
Species: Chickens (broilers, breeders and layers)
Specimens: Serum, plasma or egg yolk
Coated antigen: Purified IBV recombinant protein
Conjugate: Anti-chicken Ig-HRP conjugate (concentrated 10X)