Order Entry
China
ContactUsLinkComponent
Replacement Set for Micropipette FX 5/FX 10
Replacement Set for Micropipette FX 5/FX 10
Catalog # KART0135000
Supplier:  KARTELL
Replacement Set for Micropipette FX 5/FX 10
Catalog # KART0135000
Supplier:  KARTELL
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Set for micropipette FX 5/FX 10
  • Pk:
    1
Specifications
About this item
Spare/replacement part set for the micropipette FX 5/FX 10 series pipettors.
Set includes 1 cone, 2 PTFE seals, 2 O-rings, and 1 cone nut.