Samco™ Bio-Tite™ Narrow Mouth Specimen Container, 40 ml, 48 mm
SAMC04-0181
: SAMCO
- Cap Size:48 mm
- Sterile:-
- Version:Non sterile
- Capacity:40 ml
- Description:Specimen containers, narrow mouth, virgin polypropylene
- Cap colour:White
- Pk:600
Prevent bio-hazardous spills and conserve storage space with stackable Samco™ narrow-mouth Bio-Tite™ specimen containers. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.
- Diameter (metric): 48 mm
- Available labled or non-labled
- Dual-threaded caps create leak-resistant seal for superior safety and handling
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Virgin polypropylene allows higher clarity to see samples, and will not become brittle
- Sterile indicator tab assures tamper evidence
- Bilingual English and Spanish label facilitates accurate usage