To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Model:EVE™
- Power:100 - 240V, 50/60Hz
- Sample volume:20 µl/channel
- Description:A high throughput multi cell counter, W×L×H: 586×477×458 mm
- Environmentally Preferable:
- Cell size:5 - 80 μm
- Weight:58 kg
- Measuring range:1×10E4 - 2×10E7 cells/ml
- Pk:1
Specifications
About this item
The EVE HT™ is a high-throughput automated multiple cell counter, providing 48-sample counting in just three minutes for cell lines analyses. With small sample volume and extended capacity, the EVE HT provides accurate cell counting and viability measurements.
- Up to 48 samples at a time
- Three minutes of counting
- Only 20 μl of sample volume required
- User-independent consistency
- Staining solution trypan blue stain