Specifications
- Packed:50 slides/pack
- Description:ADAMII assay slides compatible with the ADAM counter
- Pk:50 Tests
- Slide type:AS
Specifications
About this item
ADAMII assay slide are used in the cell type of thymocytes, splenocytes, PBMCs, dendritic cells, monocytes, lymphocytes, keratinocytes, epithelial cells, established cell lines, primary cells, stem cells, blood cells, neuronal cells.
Application: Cell cycle, Apoptosis and necrosis, cell size, viability, total cell count and transfection.