Order Entry
China
ContactUsLinkComponent
J.T.Baker® BAKER BIO-ANALYZED® Buffer B: 20 mm NaOH/1,0 M NaCl in 10% Acetonitrile
J.T.Baker® BAKER BIO-ANALYZED® Buffer B: 20 mm NaOH/1,0 M NaCl in 10% Acetonitrile
Catalog # BAKR9121-51
Supplier:  Avantor
CAS Number:  
undefined
J.T.Baker® BAKER BIO-ANALYZED® Buffer B: 20 mm NaOH/1,0 M NaCl in 10% Acetonitrile
Catalog # BAKR9121-51
Supplier:  Avantor
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    20 mM NaOH/1,0 M NaCl
  • Pk:
    200 L
  • Description:
    Buffer B
  • Size:
    200 L

Specifications

About this item

  • Sodium Chloride (M): 0,95 to 1,05
  • Sodium Hydroxide (by titration) (mM): 18 to 22

Manufactured with water meeting the following requirements:

Total organic compounds (500 ppb max.)
Conductivity, (2,1 uS/cm max.)
Microbes (50 CFU max.)

Caution: For laboratory, research or manufacturing use.