Whatman™ Nuclepore Track-Etched Polycarbonate AOX Membranes, Whatman products (Cytiva)
Cytiva's Nuclepore filters are manufactured from high-quality polycarbonate film, with sharply defined pore sizes and high flow rates for reproducible AOX analysis.
- High chemical resistance and thermal stability for a wide range of samples
- Low, consistent ash levels and low tare weights
- Compatible with ISO 9562 guidelines for determining adsorbable organically bound halogens
- Smooth flat surface increases particle visibility
Explore the full range of Whatman Nuclepore polycarbonate membrane filters from Cytiva business for hydrophilic, black, and AOX membranes.
Adsorbable organic halogens (AOX) are organic compounds containing halides such as chlorine, bromine, or iodine. Some of these compounds are toxic to humans and marine life. Like mercury, AOX can concentrate up the food chain.
Specified by ISO 9562 (and EPA 1650 in the US), AOX analysis measures the quantity of halides present in water samples. The Nuclepore AOX analysis filter meets the specifications for a filter per these methods.