L(+)-Asparagine monohydrate NF, Multi-Compendial, J.T.Baker®
: Avantor
:5794-13-8
:
:C₄H₈N₂O₃·1H₂O
:MFCD00151038
:150.13 g/mol
:14,00837
:215…217 °C
:1.567 g/cm³ (20 °C)
:Ambient
:200-735-9
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- NF - Assay (dried basis) :95.5 - 102.0 %
- NF - Identification A :Passes test
- NF - Identification B :Passes test
- NF - Residue on Ignition:≤ 0.1 %
- NF - Lead (Pb):≤ 5 ppm
- NF - Chromatographic Purity: Asparagine Related Compound A :≤ 1.0 %
- NF - Chromatographic Purity: Aspartic acid:≤ 1.0 %
- NF - Chromatographic Purity: Each individual unidentified impurity:≤ 0.5 %
- NF - Chromatographic Purity: Total Impurities:≤ 3.0 %
- NF - Specific Rotation @ 20°C (+) :33.0 - 36.5 °
- NF - Total Aerobic Microbial Count (cfu/g) :≤ 1000
- NF - Total Yeast and Mold Count (cfu/g) :≤ 100
- NF - Loss on Drying at 130°C :11.5 - 12.5 %
- EP/BP - Assay (dried basis) :99.0 - 101.0 %
- EP/BP - Identification A :Passes test
- EP/BP - Identification B :Passes test
- EP/BP - Identification D :Passes test
- EP/BP - Appearance of Solution:Passes test
- EP/BP - pH:4.0 - 6.0
- EP/BP - Specific Rotation @ 20°C (+) :33.7 - 36.0 °
- EP/BP - Related Substances - Impurity A :≤ 0.5 %
- EP/BP - Related Substances - Impurity C :≤ 0.1 %
- EP/BP - Related Substances - Unspecified Impurities, each :≤ 0.05 %
- EP/BP - Related Substances - Total impurities :≤ 0.8 %
- EP/BP - Chloride (Cl) :≤ 200 ppm
- EP/BP - Sulfate (SO₄) :≤ 200 ppm
- EP/BP - Ammonium (NH₄):≤ 0.1 %
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Loss on drying (130°C):10.5 - 12.5 %
- EP/BP - Sulphated ash:≤ 0.1 %
- Endotoxin:≤ 10 EU/g