

Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Catalog # VWRC92497.320
Supplier: VWR Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay :min. 99.9%
- Water :max. 0.005%
- Evaporation Residue:max. 0.0001%
- Acidity [meq/g]:max. 0.0005
- Alkalinity [meq/g]:max. 0.00006
- Gradient use test (205 nm):max. 0.002 AU
- Gradient use test (254 nm):max. 0.0005 AU
- Baseline drift (205 nm):max. 0.02 AU
- Fluorescence (as quinine; 254 nm):max. 1 ppb
- Fluorescence (as quinine; 365 nm):max. 1 ppb
- UV transmission (190 nm):min. 18%
- UV transmission (193 nm):min. 50%
- UV transmission (197 nm):min. 80%
- UV transmission (215 nm):min. 95%
- UV transmission (230-400 nm):min. 99%
- Ag (Silver):max. 10 ppb
- Al (Aluminium):max. 25 ppb
- Ca (Calcium):max. 25 ppb
- Cu (Copper):max. 10 ppb
- Fe (Iron):max. 10 ppb
- K (Potassium):max. 10 ppb
- Mg (Magnesium):max. 10 ppb
- Mn (Manganese):max. 10 ppb
- Na (Sodium):max. 50 ppb
- Ni (Nickel):max. 10 ppb
- Pb (Lead):max. 10 ppb
- Zn (Zinc):max. 10 ppb
- Suitability for PFAS analysis:passes test
- Suitability for LC-MS:passes test
Specification Test Results
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45,7 °C
Density:0.782 g/cm³ (20 °C)
EINECS:200-835-2
UN:1648
ADR:3,II
:Packaged under nitrogen.
Flash Pt:5 °C (closed cup)
REACH:01-2119471307-38
:GHS02
:GHS07
Storage Temperature:Ambient