Specifications
- Model:NanoDrop™ One
- Communication interface:Three USB-A ports, Ethernet, Bluetooth® and Wi-Fi3
- Light source:Xenon flash lamp
- Resolution:≤1,8 nm (FWHM at Hg 254 nm), spectral bandwidth
- Sample size:1 µL (min.)
- Description:UV-Vis spectrophotometer
- Seq:001
- Path length:0,030 to 1,0 mm auto-ranging
- Wavelength range:190 - 850 nm
- Display:Built-in touchscreen
- Photometric accuracy:3% at 0,97 A, 302 nm (Abs/mm at 25 °C)
- Photometric range:(10 mm equivalent);
Pedestal: 0 – 550 A;
Cuvette: 0 – 1,5 A - Software:Windows® 7 and 10, 64 bit (PC software requirements)
- Wavelengths:Peptides (205 nm);
DNA and RNA (260 nm);
Purified protein (280 nm);
Toxicology assays and industrial dyes (490 nm);
Gold nanoparticles (520 nm);
Colorimetric protein assays (BCA 562 nm, Bradford 595 nm, Modified Lowry 650 nm, Pierce 660 660 nm);
Optical Density measurements (600 nm) - W×D×H:200×254×323 mm
- Pk:1
Specifications
About this item
Quantify and qualify DNA, RNA, and protein samples with only 1 - 2 µL and have full-spectral data in seconds using the Thermo Scientific™ NanoDrop™ One microvolume UV-Vis spectrophotometer.
- Prevent costly delays – identify contaminants, obtain corrected concentrations, and enjoy success in your downstream applications
- Collect data fast – Auto-Blank and Auto-Measure modes save time while organizing data
- Save bench space – user-friendly local control with sleek, standalone design means no PC required
- Wide spectral range (190 - 850 nm) for measuring a variety of samples types
- Pedestal measurements that require only 0.5 - 2 µL of your sample and do not require sample dilution, even for highly concentrated samples
- Pre-configured methods for DNA, Protein A280, Microarray, Protein and Labels, Pierce 660, Bradford, BCA, and Lowry
- User-friendly software that includes custom methods and data export capabilities
- Instrument comes with a 2-year warranty for worry-free support
Prevent costly delays from troubleshooting failed experiments with enhanced understanding of sample quality using the NanoDrop One spectrophotometer, built with innovative Thermo Scientific™Acclaro™ Sample Intelligence technology. Obtain accurate measurements for concentrated samples with no need for dilutions - all with the speed and simplicity of the original NanoDrop 2000 instruments you've come to rely on.
Acclaro™ technology enhances user understanding of sample quality while delivering accurate quantitative measurements. Early identification of contaminants can prevent failure of downstream applications and save days of troubleshooting. Acclaro technology allows you to verify sample quality in seconds - identify sample contaminants and obtain corrected concentration results. Access information through alerts - receive instant feedback about sample quality with on-demand technical support and guided troubleshooting. Rely on accurate results - ensure measurement integrity with embedded sensor and digital image analysis.
Ergonomic, standalone design - integrated Android™ tablet eliminates the need for a separate computer and provides modern connectivity for seamless data transfer via Wireless*, Ethernet or USB. Auto-Measure and Auto-Blank functions - multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF with a single tap on the screen.
Wider dynamic range - no dilutions required for highly concentrated samples (dsDNA at 27,500 ug/µL) with auto-range pathlength technology. Integrated Learning Center - an archive of answers with technical support documents and educational animations at your fingertips. Contains both pedestal and cuvette positions to expand experimental flexibility and increase the dynamic range. Measure dilute samples, perform kinetics experiments and take optical density measurements of bacterial cultures. Cuvette position includes temperature control and stirring.
Combining the unique sample-retention system with cuvette capability accommodates low and high concentrations (2.0 - 27,500 ng/ µL dsDNA and 0.06 - 820 mg/mL BSA).
Additional cuvette position – the Thermo Scientific™ NanoDrop™ OneC spectrophotometer adds experimental flexibility and increases the dynamic range. Use cuvettes to measure dilute samples and optical density of bacterial cultures or to perform kinetics experiments. Includes cuvette temperature control and stirring. Cuvette position can be used with instrument arm up or down.